ENA423200506 Enamine

(3,4-dihydro-2H-pyran-6-yl)methanol

  • MDL number MFCD20126379
  • Cas Number : 72081-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-104032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-104032
Availability details for
Quantity
  • Availability
  • Add to Cart