ENA423200508 Enamine

2-methoxy-4-methyl-6-(prop-2-en-1-yl)phenol

  • MDL number MFCD11500864
  • Cas Number : 77879-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159897
Availability details for
Quantity
  • Availability
  • Add to Cart