ENA423200513 Enamine

1,2,3,4,4a,9,9a,10-octahydroacridine

  • MDL number MFCD20661281
  • Cas Number : 92039-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159920
Availability details for
Quantity
  • Availability
  • Add to Cart