ENA423200516 Enamine

2-methyl-1-(2-methylphenyl)propan-1-ol

  • MDL number MFCD12153670
  • Cas Number : 100994-03-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-159925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-159925
Availability details for
Quantity
  • Availability
  • Add to Cart