ENA423200524 Enamine

4H,5H,6H-cyclopenta[b]thiophen-6-one

  • MDL number MFCD03426936
  • Cas Number : 5650-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57...59
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-136799

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-136799
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart