ENA423200531 Enamine

2-chloro-3-(ethanesulfonyl)pyridine

  • MDL number MFCD00661251
  • Cas Number : 87695-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-137712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-137712
Availability details for
Quantity
  • Availability
  • Add to Cart