ENA423200551 Enamine

6-chloro-5-nitropyridine-3-carbonitrile

  • MDL number MFCD18905778
  • Cas Number : 160906-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-139033
Availability details for
Quantity
  • Availability
  • Add to Cart