ENA423200552 Enamine

furan-2-sulfonyl chloride

  • MDL number MFCD06658968
  • Cas Number : 52665-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139041

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139041
Availability details for
Quantity
  • Availability
  • Add to Cart