ENA423200566 Enamine

(benzyloxy)(but-3-en-1-yl)amine

  • MDL number MFCD21116692
  • Cas Number : 1489935-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160057

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160057
Availability details for
Quantity
  • Availability
  • Add to Cart