ENA423200575 Enamine

3-methylthieno[3,2-b]pyridine-6-carboxylic acid

  • MDL number MFCD27980726
  • Cas Number : 1372924-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160072

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160072
Availability details for
Quantity
  • Availability
  • Add to Cart