ENA423200578 Enamine

2-tert-butyl-4-fluoro-5-methylphenol

  • MDL number MFCD24713798
  • Cas Number : 220901-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160082

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160082
Availability details for
Quantity
  • Availability
  • Add to Cart