ENA423200579 Enamine

2-(dimethyl-4H-1,2,4-triazol-3-yl)acetonitrile

  • MDL number MFCD27959437
  • Cas Number : 1598277-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160084

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160084
Availability details for
Quantity
  • Availability
  • Add to Cart