ENA423200580 Enamine

5-phenyl-1,3,4-thiadiazole-2-carbonitrile

  • MDL number MFCD27959438
  • Cas Number : 70391-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160085

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160085
Availability details for
Quantity
  • Availability
  • Add to Cart