ENA423200585 Enamine

4-(bromomethyl)-3-chlorobenzonitrile

  • MDL number MFCD01208294
  • Cas Number : 21924-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160097

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-160097
pictograms GHS05,GHS08
Availability details for
Quantity
  • Availability
  • Add to Cart