ENA423200590 Enamine

7-nitroquinolin-4-ol

  • MDL number MFCD08688614
  • Cas Number : 6270-14-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 317-320
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160122
Availability details for
Quantity
  • Availability
  • Add to Cart