ENA423200597 Enamine

1-methyl-1,4-diazepan-2-one hydrochloride

  • MDL number MFCD19443962
  • Cas Number : 685859-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-160143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160143
Availability details for
Quantity
  • Availability
  • Add to Cart