ENA423200599 Enamine

6-chloro-2-methylimidazo[1,2-a]pyridin-3-amine

  • MDL number MFCD14540395
  • Cas Number : 1215970-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160165
Availability details for
Quantity
  • Availability
  • Add to Cart