ENA423200600 Enamine

3-methoxypropane-1,2-diamine

  • MDL number MFCD19204176
  • Cas Number : 101857-37-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160167
Availability details for
Quantity
  • Availability
  • Add to Cart