ENA423200603 Enamine

N-(2-amino-4-methoxyphenyl)naphthalene-1-carboxamide

  • MDL number MFCD09814391
  • Cas Number : 1016758-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139398

Safety Information

msds https://www.enaminestore.com/catalog/EN300-139398
Availability details for
Quantity
  • Availability
  • Add to Cart