ENA423200609 Enamine

(5-bromothiophen-2-yl)(cyclopropyl)methanone

  • MDL number MFCD12786026
  • Cas Number : 1216229-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160176

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160176
Availability details for
Quantity
  • Availability
  • Add to Cart