ENA423200611 Enamine

4-[6-(bromomethyl)-5H,6H-[1,2,4]triazolo[3,4-b][1,3]thiazol-3-yl]pyridine

  • MDL number MFCD04188276
  • Cas Number : 848207-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-160185

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160185
Availability details for
Quantity
  • Availability
  • Add to Cart