ENA423200622 Enamine

4-bromoquinolin-8-ol

  • MDL number MFCD08688609
  • Cas Number : 139399-63-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-139995

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-139995
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart