ENA423200625 Enamine

5-({[(tert-butoxy)carbonyl]amino}methyl)furan-3-carboxylic acid

  • MDL number MFCD27980629
  • Cas Number : 1797772-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140085

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140085
Availability details for
Quantity
  • Availability
  • Add to Cart