ENA423200628 Enamine

(4-amino-2,2,3-trimethylcyclohexyl)methanol

  • MDL number MFCD09035034
  • Cas Number : 1053170-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160253

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160253
Availability details for
Quantity
  • Availability
  • Add to Cart