ENA423200635 Enamine

1-(5-methyl-1H-pyrazol-3-yl)piperazine

  • MDL number MFCD22394065
  • Cas Number : 1491491-75-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-160273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160273
Availability details for
Quantity
  • Availability
  • Add to Cart