ENA423200644 Enamine

1-(5-methyl-4H-1,2,4-triazol-3-yl)piperazine

  • MDL number MFCD27980631
  • Cas Number : 111641-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-140416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140416
Availability details for
Quantity
  • Availability
  • Add to Cart