ENA423200645 Enamine

ethyl 2-(2-phenylethenyl)-1,3-oxazole-4-carboxylate

  • Cas Number : 107367-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-140588

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-140588
Availability details for
Quantity
  • Availability
  • Add to Cart