ENA423200647 Enamine

(5-bromothiophen-2-yl)(cyclopropyl)methanol

  • MDL number MFCD12788271
  • Cas Number : 1216254-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160289
Availability details for
Quantity
  • Availability
  • Add to Cart