ENA423200653 Enamine

cyclohexyl(phenyl)methanol

  • MDL number MFCD00019357
  • Cas Number : 945-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-160298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160298
Availability details for
Quantity
  • Availability
  • Add to Cart