ENA423200655 Enamine

N'-[amino(pyrazin-2-yl)methylidene]acetohydrazide

  • MDL number MFCD05721808
  • Cas Number : 717846-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160301

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160301
Availability details for
Quantity
  • Availability
  • Add to Cart