ENA423200658 Enamine

3-nitroquinolin-2-ol

  • MDL number MFCD09754108
  • Cas Number : 103029-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160346

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160346
Availability details for
Quantity
  • Availability
  • Add to Cart