ENA423200661 Enamine

6-bromo-3-methylpyridine-2-carbonitrile

  • MDL number MFCD19707627
  • Cas Number : 1379335-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160352
Availability details for
Quantity
  • Availability
  • Add to Cart