ENA423200664 Enamine

2,2-dimethyl-3-[4-(trifluoromethyl)phenyl]propanoic acid

  • MDL number MFCD17525708
  • Cas Number : 1439896-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160357
Availability details for
Quantity
  • Availability
  • Add to Cart