ENA423200667 Enamine

3,5-dinitro-N-(pyridin-3-ylmethyl)benzamide

  • MDL number MFCD00495413
  • Cas Number : 58624-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-140947

Safety Information

msds https://www.enaminestore.com/catalog/EN300-140947
Availability details for
Quantity
  • Availability
  • Add to Cart