ENA423200669 Enamine

3-(4-aminophenyl)-4,5-dihydro-1,2,4-oxadiazol-5-one hydrochloride

  • MDL number MFCD16295161
  • Cas Number : 1341040-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160366

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-160366
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart