ENA423200670 Enamine

tert-butyl N-[3-(4-methylpiperazin-1-yl)-3-oxopropyl]carbamate

  • MDL number MFCD27959456
  • Cas Number : 1616684-41-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-160378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160378
Availability details for
Quantity
  • Availability
  • Add to Cart