ENA423200681 Enamine

3-ethylcyclohex-2-en-1-one

  • MDL number MFCD20882082
  • Cas Number : 17299-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160472
Availability details for
Quantity
  • Availability
  • Add to Cart