ENA423200682 Enamine

4H,5H,6H,7H-[1,2,3]triazolo[1,5-a]pyridine

  • MDL number MFCD13176208
  • Cas Number : 210880-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160473
Availability details for
Quantity
  • Availability
  • Add to Cart