ENA423200683 Enamine

2-(benzyloxy)-4-methoxyaniline

  • MDL number MFCD24977842
  • Cas Number : 807266-13-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160480

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160480
Availability details for
Quantity
  • Availability
  • Add to Cart