ENA423200686 Enamine

N,N-dimethyl-5-(piperidin-2-yl)-1,2,4-oxadiazol-3-amine

  • MDL number MFCD27959460
  • Cas Number : 1597268-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160483

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160483
Availability details for
Quantity
  • Availability
  • Add to Cart