ENA423200689 Enamine

2-{1,4-dioxaspiro[4.5]decan-8-yl}ethan-1-ol

  • MDL number MFCD07779436
  • Cas Number : 135761-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141284

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-141284
Availability details for
Quantity
  • Availability
  • Add to Cart