ENA423200691 Enamine

1-(benzylamino)-2-methylbutan-2-ol

  • MDL number MFCD09260639
  • Cas Number : 939793-33-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160489

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160489
Availability details for
Quantity
  • Availability
  • Add to Cart