ENA423200695 Enamine

5-methoxy-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid

  • MDL number MFCD20647678
  • Cas Number : 6944-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160498
Availability details for
Quantity
  • Availability
  • Add to Cart