ENA423200696 Enamine

tert-butyl 2-(2-hydroxyethyl)morpholine-4-carboxylate

  • MDL number MFCD09608014
  • Cas Number : 913642-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160501

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160501
Availability details for
Quantity
  • Availability
  • Add to Cart