ENA423200700 Enamine

3-methyl-2-(1-methyl-1H-pyrazol-5-yl)butan-1-amine

  • MDL number MFCD27980634
  • Cas Number : 1522545-56-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141482

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141482
Availability details for
Quantity
  • Availability
  • Add to Cart