ENA423200701 Enamine

1-{6-[(2,2,2-trifluoroethoxy)methyl]pyridin-3-yl}methanamine

  • MDL number MFCD23844626
  • Cas Number : 1513116-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 73
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160510

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160510
Availability details for
Quantity
  • Availability
  • Add to Cart