ENA423200703 Enamine

2,2,2-trifluoro-1-(4-methyl-1H-indol-3-yl)ethan-1-one

  • MDL number MFCD24107294
  • Cas Number : 79878-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160536

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160536
Availability details for
Quantity
  • Availability
  • Add to Cart