ENA423200709 Enamine

1,5-dimethyl-3-nitro-1H-pyrazole

  • MDL number MFCD04971109
  • Cas Number : 59376-16-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-141534

Safety Information

msds https://www.enaminestore.com/catalog/EN300-141534
Availability details for
Quantity
  • Availability
  • Add to Cart