ENA423200712 Enamine

6-amino-1-(methoxymethyl)-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD21223085
  • Cas Number : 1519406-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160599
Availability details for
Quantity
  • Availability
  • Add to Cart