ENA423200714 Enamine

2-chloro-N-(2-hydroxy-2-methylpropyl)acetamide

  • MDL number MFCD21126395
  • Cas Number : 1477831-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160602
Availability details for
Quantity
  • Availability
  • Add to Cart